2,3-Dichloro-5-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

62835-500MG 62835-1G 62835-25G 62835-5G
77331-786EA 301.79 USD
77331-786 77356-044 77389-270 77380-104
2,3-Dichloro-5-methoxybenzaldehyde ≥97%
2,3-Dichloro-5-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆Cl₂O₂
MW: 205.0380 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761064
CAS Number: 2090464-85-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR