3-Iodo-5-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

62827-500MG 62827-1G 62827-5G 62827-25G
77331-784EA 206 USD
77331-784 77356-042 77367-916 77389-268
3-Iodo-5-methoxybenzaldehyde ≥97%
3-Iodo-5-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₇IO₂
Storage Temperature: Ambient
MDL Number: MFCD22544341
CAS Number: 152734-38-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR