2-Fluoro-4-iodo-3-methoxy-1-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

62818-500MG 62818-1G 62818-5G
77337-764EA 440.84 USD
77337-764 77356-034 77380-098
2-Fluoro-4-iodo-3-methoxy-1-methylbenzene ≥95%
2-Fluoro-4-iodo-3-methoxy-1-methylbenzene

Precaución: for Research Use Only

Formula: C₈H₈FIO
Storage Temperature: Ambient
CAS Number: 2386931-95-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR