(2,5-Difluoro-4-methoxyphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

62817-500MG 62817-1G 62817-5G 62817-25G
77331-778EA 115.36 USD
77331-778 77343-402 77367-912 77380-096
(2,5-Difluoro-4-methoxyphenyl)(methyl)sulfane ≥97%
(2,5-Difluoro-4-methoxyphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₈H₈F₂OS
Storage Temperature: Ambient
MDL Number: MFCD28503408
CAS Number: 1934690-72-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR