1-Ethyl-2,5-difluoro-4-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

62816-500MG 62816-1G 62816-5G
77336-846EA 394.49 USD
77336-846 77356-032 77380-094
1-Ethyl-2,5-difluoro-4-methoxybenzene ≥97%
1-Ethyl-2,5-difluoro-4-methoxybenzene

Precaución: for Research Use Only

Formula: C₉H₁₀F₂O
Storage Temperature: Ambient
MDL Number: MFCD34761056
CAS Number: 1893194-72-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR