2,4-Difluoro-3-methyl-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

62788-500MG 62788-1G 62788-5G 62788-25G
77331-772EA 150.38 USD
77331-772 77345-364 77367-904 77389-262
2,4-Difluoro-3-methyl-1,1'-biphenyl ≥97%
2,4-Difluoro-3-methyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₁₀F₂
MW: 204.2153 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761041
CAS Number: 76350-72-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR