Methyl 2,4-difluoro-4'-methyl-[1,1'-biphenyl]-3-carboxylate ≥97%

Supplier: AOBChem USA

Warning

62783-500MG 62783-1G 62783-5G
77331-764EA 340.93 USD
77331-764 77356-024 77380-082
Methyl 2,4-difluoro-4'-methyl-[1,1'-biphenyl]-3-carboxylate ≥97%
Methyl 2,4-difluoro-4'-methyl-[1,1'-biphenyl]-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₅H₁₂F₂O₂
MW: 262.2513 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761037
CAS Number: 2901082-01-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR