3-(Difluoromethoxy)benzyl alcohol ≥95%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
(3-(Difluoromethoxy)phenyl)methanol
62774-500MG
62774-1G
62774-5G
62774-25G
77331-760EA
136.99
USD
77331-760
77344-608
77367-896
77389-254
3-(Difluoromethoxy)benzyl alcohol ≥95%
3-(Difluoromethoxy)benzyl alcohol
Precaución: for Research Use Only
Formula:
C₈H₈F₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD00236227 CAS Number: 125903-81-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...