Methyl 3-(difluoromethoxy)benzoate ≥97%

Supplier: AOBChem USA

Warning

62772-500MG 62772-1G 62772-5G 62772-25G
77331-756EA 204.97 USD
77331-756 77356-018 77367-892 77389-250
Methyl 3-(difluoromethoxy)benzoate ≥97%
Methyl 3-(difluoromethoxy)benzoate

Precaución: for Research Use Only

Formula: C₉H₈F₂O₃
MW: 202.1548 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06203752
CAS Number: 773873-87-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR