4-Chloro-2-ethoxy-5-fluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

62757-500MG 62757-1G 62757-5G
77336-114EA 375.95 USD
77336-114 77356-012 77380-074
4-Chloro-2-ethoxy-5-fluorobenzaldehyde ≥97%
4-Chloro-2-ethoxy-5-fluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₈ClFO₂
Storage Temperature: Ambient
MDL Number: MFCD06246182
CAS Number: 883521-86-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR