4-Chloro-2-ethoxy-5-fluorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

62755-500MG 62755-1G 62755-5G
77337-600EA 425.39 USD
77337-600 77356-010 77380-072
4-Chloro-2-ethoxy-5-fluorobenzoic acid ≥97%
4-Chloro-2-ethoxy-5-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₉H₈ClFO₃
MW: 218.6094 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28627844
CAS Number: 1784105-20-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR