(2-Chloro-4-ethoxy-5-methylphenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

62747-500MG 62747-1G 62747-5G
77337-822EA 448.05 USD
77337-822 77356-002 77380-064
(2-Chloro-4-ethoxy-5-methylphenyl)boronic acid ≥97%
(2-Chloro-4-ethoxy-5-methylphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₉H₁₂BClO₃
MW: 214.4537 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23380963
CAS Number: 1393536-47-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR