Methyl 2-chloro-4-ethoxy-5-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

62745-500MG 62745-1G 62745-5G
77336-648EA 390.37 USD
77336-648 77356-000 77380-062
Methyl 2-chloro-4-ethoxy-5-methylbenzoate ≥97%
Methyl 2-chloro-4-ethoxy-5-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₃ClO₃
MW: 228.6721 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761020
CAS Number: 2384353-21-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR