2-Chloro-4-isopropoxy-5-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

62738-500MG 62738-1G 62738-5G
77340-298EA 895.07 USD
77340-298 77355-994 77380-056
2-Chloro-4-isopropoxy-5-methylbenzoic acid ≥97%
2-Chloro-4-isopropoxy-5-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₃ClO₃
Storage Temperature: Ambient
MDL Number: MFCD34761014
CAS Number: 2807465-20-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR