Methyl 2-ethoxy-5-ethylbenzoate ≥97%

Supplier: AOBChem USA

Warning

62737-500MG 62737-1G 62737-5G 62737-25G
77331-754EA 225.57 USD
77331-754 77355-992 77367-890 77389-248
Methyl 2-ethoxy-5-ethylbenzoate ≥97%
Methyl 2-ethoxy-5-ethylbenzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₃
MW: 208.2536 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31773587
CAS Number: 1367990-75-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR