4-Methoxy-2,3-dimethyl-phenol ≥97%

Supplier: AOBChem USA

Warning

62731-500MG 62731-1G 62731-5G 62731-25G
77331-750EA 224.54 USD
77331-750 77355-988 77367-888 77389-246
4-Methoxy-2,3-dimethyl-phenol ≥97%
4-Methoxy-2,3-dimethyl-phenol

Precaución: for Research Use Only

Formula: C₉H₁₂O₂
Storage Temperature: Ambient
MDL Number: MFCD09032519
CAS Number: 35355-33-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR