4-(Benzyloxy)-2-chloro-5-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

62724-500MG 62724-1G 62724-5G
77340-152EA 796.19 USD
77340-152 77355-974 77380-040
4-(Benzyloxy)-2-chloro-5-methylbenzoic acid ≥97%
4-(Benzyloxy)-2-chloro-5-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₃ClO₃
Storage Temperature: Ambient
MDL Number: MFCD34761007
CAS Number: 2764731-91-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR