3-Chloro-4-fluoro-5-formylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

62705-500MG 62705-1G 62705-5G
77336-112EA 375.95 USD
77336-112 77355-970 77380-038
3-Chloro-4-fluoro-5-formylbenzoic acid ≥97%
3-Chloro-4-fluoro-5-formylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₄ClFO₃
MW: 202.5669 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760994
CAS Number: 2385884-08-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR