5-(2-Aminothiazol-5-yl)-2-methylphenol ≥95%

Supplier: AOBChem USA

Warning

62701-500MG 62701-1G 62701-5G
77331-744EA 342.99 USD
77331-744 77355-968 77380-034
5-(2-Aminothiazol-5-yl)-2-methylphenol ≥95%
5-(2-Aminothiazol-5-yl)-2-methylphenol

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂OS
Storage Temperature: Ambient
CAS Number: 2807450-29-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR