5-(5-Chloro-2-fluoro-3-(trifluoromethyl)phenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

62700-500MG 62700-1G 62700-5G
77336-524EA 388.31 USD
77336-524 77355-966 77380-032
5-(5-Chloro-2-fluoro-3-(trifluoromethyl)phenyl)thiazol-2-amine ≥97%
5-(5-Chloro-2-fluoro-3-(trifluoromethyl)phenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₅ClF₄N₂S
MW: 296.6717 g/mol
Storage Temperature: Ambient
CAS Number: 2807441-68-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR