5-(3,4-Difluoro-2-methoxyphenyl)thiazol-2-amine ≥95%

Supplier: AOBChem USA

Warning

62698-500MG 62698-1G 62698-5G
77337-110EA 407.88 USD
77337-110 77355-964 77380-030
5-(3,4-Difluoro-2-methoxyphenyl)thiazol-2-amine ≥95%
5-(3,4-Difluoro-2-methoxyphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₈F₂N₂OS
MW: 242.2451 g/mol
Storage Temperature: Ambient
CAS Number: 2764733-91-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR