5-(3,4-Difluoro-2-methylphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

62696-500MG 62696-1G 62696-5G
77339-598EA 602.55 USD
77339-598 77355-960 77380-026
5-(3,4-Difluoro-2-methylphenyl)thiazol-2-amine ≥97%
5-(3,4-Difluoro-2-methylphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₈F₂N₂S
MW: 226.2457 g/mol
Storage Temperature: Ambient
CAS Number: 2901065-98-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR