3-Acetyl-4-fluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

62686-500MG 62686-1G 62686-5G
77336-256EA 384.19 USD
77336-256 77355-958 77380-024
3-Acetyl-4-fluorobenzaldehyde ≥97%
3-Acetyl-4-fluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₇FO₂
MW: 166.1490 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17015608
CAS Number: 319455-11-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR