3-Acetyl-4-fluorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

62684-500MG 62684-1G 62684-5G 62684-25G
77331-740EA 204.97 USD
77331-740 77355-956 77367-882 77389-240
3-Acetyl-4-fluorobenzoic acid ≥97%
3-Acetyl-4-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₉H₇FO₃
MW: 182.148 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22493249
CAS Number: 1505662-44-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR