2-Chloro-6-iodo-3-(trifluoromethyl)phenol ≥97%

Supplier: AOBChem USA

Warning

62681-500MG 62681-1G 62681-5G
77331-736EA 318.27 USD
77331-736 77355-954 77380-022
2-Chloro-6-iodo-3-(trifluoromethyl)phenol ≥97%
2-Chloro-6-iodo-3-(trifluoromethyl)phenol

Precaución: for Research Use Only

Formula: C₇H₃ClF₃IO
MW: 322.4507 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760976
CAS Number: 2386558-17-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR