2-Chloro-4-iodo-3-(methoxymethoxy)-1-(trifluoromethyl)benzene ≥95%

Supplier: AOBChem USA

Warning

62680-500MG 62680-1G 62680-5G 62680-25G
77331-734EA 121.54 USD
77331-734 77343-964 77367-878 77380-020
2-Chloro-4-iodo-3-(methoxymethoxy)-1-(trifluoromethyl)benzene ≥95%
2-Chloro-4-iodo-3-(methoxymethoxy)-1-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₉H₇ClF₃IO₂
MW: 366.5033 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760975
CAS Number: 2807465-59-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR