3-Chloro-2-hydroxy-4-(trifluoromethyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

62677-500MG 62677-1G 62677-5G 62677-25G
77331-730EA 233.81 USD
77331-730 77355-952 77367-874 77389-236
3-Chloro-2-hydroxy-4-(trifluoromethyl)benzoic acid ≥97%
3-Chloro-2-hydroxy-4-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₄ClF₃O₃
Storage Temperature: Ambient
MDL Number: MFCD34760972
CAS Number: 2383771-33-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR