3-Chloro-2-hydroxy-4-(trifluoromethyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

62674-500MG 62674-1G 62674-5G
77331-728EA 342.99 USD
77331-728 77355-950 77380-016
3-Chloro-2-hydroxy-4-(trifluoromethyl)benzaldehyde ≥97%
3-Chloro-2-hydroxy-4-(trifluoromethyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₈H₄ClF₃O₂
MW: 224.5643 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760970
CAS Number: 2807445-43-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR