(3-(Trifluoromethyl)-1,2-phenylene)bis(methylsulfane) ≥97%

Supplier: AOBChem USA

Warning

62662-500MG 62662-1G 62662-5G 62662-25G
77331-724EA 122.57 USD
77331-724 77343-962 77367-864 77380-004
(3-(Trifluoromethyl)-1,2-phenylene)bis(methylsulfane) ≥97%
(3-(Trifluoromethyl)-1,2-phenylene)bis(methylsulfane)

Precaución: for Research Use Only

Formula: C₉H₉F₃S₂
MW: 238.2929 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760964
CAS Number: 2764733-85-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR