3-Chloro-4-((4-fluorobenzyl)oxy)phenol ≥97%

Supplier: AOBChem USA

Warning

62661-500MG 62661-1G 62661-5G 62661-25G
77331-722EA 110.21 USD
77331-722 77343-242 77367-862 77380-002
3-Chloro-4-((4-fluorobenzyl)oxy)phenol ≥97%
3-Chloro-4-((4-fluorobenzyl)oxy)phenol

Precaución: for Research Use Only

Formula: C₁₃H₁₀ClFO₂
MW: 252.6687 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760963
CAS Number: 2901083-27-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR