Ethyl 3-chloro-4-((4-fluorobenzyl)oxy)benzoate ≥97%

Supplier: AOBChem USA

Warning

62658-500MG 62658-1G 62658-5G 62658-25G
77331-720EA 236.9 USD
77331-720 77355-946 77367-858 77389-232
Ethyl 3-chloro-4-((4-fluorobenzyl)oxy)benzoate ≥97%
Ethyl 3-chloro-4-((4-fluorobenzyl)oxy)benzoate

Precaución: for Research Use Only

Formula: C₁₆H₁₄ClFO₃
MW: 308.7319 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28102249
CAS Number: 1706450-49-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR