3-Chloro-4-((4-fluorobenzyl)oxy)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

62655-500MG 62655-1G 62655-5G 62655-25G
77331-716EA 225.57 USD
77331-716 77355-942 77367-854 77389-228
3-Chloro-4-((4-fluorobenzyl)oxy)benzaldehyde ≥97%
3-Chloro-4-((4-fluorobenzyl)oxy)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₄H₁₀ClFO₂
MW: 264.6794 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03030597
CAS Number: 433250-87-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR