4-Ethoxy-2-fluoro-5-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

62651-500MG 62651-1G 62651-5G
77338-878EA 527.36 USD
77338-878 77355-940 77379-998
4-Ethoxy-2-fluoro-5-methylbenzoic acid ≥97%
4-Ethoxy-2-fluoro-5-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁FO₃
MW: 198.1909 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760959
CAS Number: 2091420-92-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR