4-Bromo-2,5-dichloro-4'-fluoro-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

62641-500MG 62641-1G 62641-5G 62641-25G
77331-708EA 85.49 USD
77331-708 77342-254 77367-846 77379-992
4-Bromo-2,5-dichloro-4'-fluoro-1,1'-biphenyl ≥97%
4-Bromo-2,5-dichloro-4'-fluoro-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₂H₆BrCl₂F
MW: 319.9844 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760951
CAS Number: 2901065-91-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR