4'-Bromo-2',5'-dichloro-[1,1'-biphenyl]-4-carbaldehyde ≥95%

Supplier: AOBChem USA

Warning

62639-500MG 62639-1G 62639-5G 62639-25G
77331-706EA 84.46 USD
77331-706 77342-206 77367-844 77379-990
4'-Bromo-2',5'-dichloro-[1,1'-biphenyl]-4-carbaldehyde ≥95%
4'-Bromo-2',5'-dichloro-[1,1'-biphenyl]-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₇BrCl₂O
MW: 330.0040 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760950
CAS Number: 2807456-53-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR