(4'-Ethyl-3-fluoro-[1,1'-biphenyl]-4-yl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

62634-500MG 62634-1G 62634-5G
77340-392EA 998.07 USD
77340-392 77355-934 77379-988
(4'-Ethyl-3-fluoro-[1,1'-biphenyl]-4-yl)boronic acid ≥97%
(4'-Ethyl-3-fluoro-[1,1'-biphenyl]-4-yl)boronic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₄BFO₂
MW: 244.0691 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10687183
CAS Number: 900796-46-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR