4-Bromo-4'-ethyl-3-fluoro-1,1'-biphenyl ≥95%

Supplier: AOBChem USA

Warning

62633-500MG 62633-1G 62633-5G
77335-776EA 365.65 USD
77335-776 77355-932 77379-986
4-Bromo-4'-ethyl-3-fluoro-1,1'-biphenyl ≥95%
4-Bromo-4'-ethyl-3-fluoro-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₂BrF
MW: 279.1474 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10687182
CAS Number: 1027512-45-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR