3-Bromo-2-ethoxy-5-fluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

62617-500MG 62617-1G 62617-5G
77336-014EA 373.89 USD
77336-014 77355-930 77379-984
3-Bromo-2-ethoxy-5-fluorobenzaldehyde ≥97%
3-Bromo-2-ethoxy-5-fluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₈BrFO₂
MW: 247.0610 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28396358
CAS Number: 1784109-49-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR