3,4-Difluoro-2-formylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

62608-500MG 62608-1G 62608-5G
77331-702EA 345.05 USD
77331-702 77355-926 77379-982
3,4-Difluoro-2-formylbenzoic acid ≥97%
3,4-Difluoro-2-formylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₄F₂O₃
MW: 186.1123 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760930
CAS Number: 2385273-71-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR