(3,4-Dibromo-2-fluorophenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

62603-500MG 62603-1G 62603-5G 62603-25G
77331-700EA 127.72 USD
77331-700 77344-228 77367-840 77379-978
(3,4-Dibromo-2-fluorophenyl)(methyl)sulfane ≥97%
(3,4-Dibromo-2-fluorophenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₇H₅Br₂FS
MW: 299.9860 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28784958
CAS Number: 1806346-45-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR