1-Fluoro-2-isopropoxy-4,5-dimethoxybenzene ≥97%

Supplier: AOBChem USA

Warning

62586-500MG 62586-1G 62586-5G
77339-694EA 614.91 USD
77339-694 77355-922 77379-976
1-Fluoro-2-isopropoxy-4,5-dimethoxybenzene ≥97%
1-Fluoro-2-isopropoxy-4,5-dimethoxybenzene

Precaución: for Research Use Only

Formula: C₁₁H₁₅FO₃
MW: 214.2334 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760918
CAS Number: 2901103-45-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR