1-(Benzyloxy)-2-fluoro-4,5-dimethoxybenzene ≥95%

Supplier: AOBChem USA

Warning

62585-500MG 62585-1G 62585-5G
77339-558EA 596.37 USD
77339-558 77355-920 77379-974
1-(Benzyloxy)-2-fluoro-4,5-dimethoxybenzene ≥95%
1-(Benzyloxy)-2-fluoro-4,5-dimethoxybenzene

Precaución: for Research Use Only

Formula: C₁₅H₁₅FO₃
MW: 262.2762 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760917
CAS Number: 2901101-76-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR