(4-Bromo-5-chloro-2-fluorophenyl)boronic acid ≥95%

Supplier: AOBChem USA

Warning

62573-500MG 62573-1G 62573-5G
77331-698EA 322.39 USD
77331-698 77355-916 77379-970
(4-Bromo-5-chloro-2-fluorophenyl)boronic acid ≥95%
(4-Bromo-5-chloro-2-fluorophenyl)boronic acid

Precaución: for Research Use Only

Formula: C₆H₄BBrClFO₂
MW: 253.2611 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760915
CAS Number: 2795134-22-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR