1-Bromo-3-ethoxy-5-(methylsulfonyl)benzene ≥97%

Supplier: AOBChem USA

Warning

62563-500MG 62563-1G 62563-5G 62563-25G
77331-696EA 97.85 USD
77331-696 77342-678 77367-836 77379-966
1-Bromo-3-ethoxy-5-(methylsulfonyl)benzene ≥97%
1-Bromo-3-ethoxy-5-(methylsulfonyl)benzene

Precaución: for Research Use Only

Formula: C₉H₁₁BrO₃S
MW: 279.1508 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760907
CAS Number: 1628604-79-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR