(3-Ethoxy-5-iodophenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

62562-500MG 62562-1G 62562-5G 62562-25G
77331-694EA 209.09 USD
77331-694 77355-914 77367-834 77389-220
(3-Ethoxy-5-iodophenyl)(methyl)sulfane ≥97%
(3-Ethoxy-5-iodophenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₉H₁₁IOS
Storage Temperature: Ambient
MDL Number: MFCD34760906
CAS Number: 2901102-12-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR