(3-Bromo-5-ethoxyphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

62554-1G 62554-5G 62554-25G
77341-882EA 106.09 USD
77341-882 77367-830 77379-964
(3-Bromo-5-ethoxyphenyl)(methyl)sulfane ≥97%
(3-Bromo-5-ethoxyphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₉H₁₁BrOS
Storage Temperature: Ambient
MDL Number: MFCD34760898
CAS Number: 2764729-04-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR