(3-Iodo-5-methoxyphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

62551-1G 62551-5G 62551-25G
77341-698EA 100.94 USD
77341-698 77367-828 77379-962
(3-Iodo-5-methoxyphenyl)(methyl)sulfane ≥97%
(3-Iodo-5-methoxyphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₈H₉IOS
MW: 280.1259 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22489305
CAS Number: 1824587-89-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR