3-Methoxy-5-(methylthio)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

62547-500MG 62547-1G 62547-5G
77331-688EA 315.18 USD
77331-688 77355-910 77379-958
3-Methoxy-5-(methylthio)benzaldehyde ≥97%
3-Methoxy-5-(methylthio)benzaldehyde

Precaución: for Research Use Only

Formula: C₉H₁₀O₂S
MW: 182.2395 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29122099
CAS Number: 1289031-82-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR