(3-Bromo-5-methoxyphenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

62542-1G 62542-5G 62542-25G
77340-790EA 60.77 USD
77340-790 77367-822 77379-956
(3-Bromo-5-methoxyphenyl)(methyl)sulfane ≥95%
(3-Bromo-5-methoxyphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₈H₉BrOS
Storage Temperature: Ambient
MDL Number: MFCD22489304
CAS Number: 1823025-16-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR