5-(3-(Benzyloxy)-4-methoxyphenyl)thiazol-2-amine ≥95%

Supplier: AOBChem USA

Warning

62526-500MG 62526-1G 62526-25G 62526-5G
77331-684EA 301.79 USD
77331-684 77355-908 77389-214 77379-954
5-(3-(Benzyloxy)-4-methoxyphenyl)thiazol-2-amine ≥95%
5-(3-(Benzyloxy)-4-methoxyphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₇H₁₆N₂O₂S
MW: 312.3861 g/mol
Storage Temperature: Ambient
CAS Number: 2807450-23-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR